AMBH580646ED Ambeed, Inc.

6-Chloro-2-(trifluoromethyl)pyrimidin-4-amine

  • MDL number MFCD08436599
  • Cas Number : 1480-66-6
  • Molecular Weight : 197.5456

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,2-8
Physical Form solid
Shipping Temperature Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1480-66-6.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338-P310
MSDS https://www.ambeed.com/products/1480-66-6.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart