AMBH580646EE Ambeed, Inc.

2-(7-Chloro-1H-indol-3-yl)acetic acid

  • MDL number MFCD09751728
  • Cas Number : 1912-41-0
  • Molecular Weight : 209.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Flash Point N/A
Storage temp. Keep in dark place,Sealed in dry,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/1912-41-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1912-41-0.html
pictograms GHS07
hstatements H319-H315-H335
Availability details for
Quantity
  • Availability
  • Add to Cart