AMBH580646F2 Ambeed, Inc.

6-Phenyl-1H-indazole

  • MDL number MFCD11109933
  • Cas Number : 1260897-38-8
  • Molecular Weight : 194.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-phenyl-1H-indazole
Inchi Code 1S/C13H10N2/c1-2-4-10(5-3-1)11-6-7-12-9-14-15-13(12)8-11/h1-9H,(H,14,15)
Inchi Key OPJICVPEFWXDDJ-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Keep in dark place,Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/1260897-38-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1260897-38-8.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart