AMBH580646F8 Ambeed, Inc.

Chloro(crotyl)(tri-tert-butylphosphine)palladium(II)

  • MDL number MFCD25372547
  • Cas Number : 1334497-00-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.ambeed.com/products/1334497-00-5.html

Safety Information

MSDS https://www.ambeed.com/products/1334497-00-5.html
Availability details for
Quantity
  • Availability
  • Add to Cart