AMBH580646F9 Ambeed, Inc.

H-?-Glu-Cys-OH

  • MDL number MFCD00237863
  • Cas Number : 636-58-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.ambeed.com/products/636-58-8.html

Safety Information

MSDS https://www.ambeed.com/products/636-58-8.html
Availability details for
Quantity
  • Availability
  • Add to Cart