AMBH580646FA Ambeed, Inc.

Delafloxacin meglumine

  • MDL number MFCD26142921
  • Cas Number : 352458-37-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Room Temp
Physical Form solid
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.ambeed.com/products/352458-37-8.html

Safety Information

MSDS https://www.ambeed.com/products/352458-37-8.html
Availability details for
Quantity
  • Availability
  • Add to Cart