AMBH580646FF Ambeed, Inc.

ML390

  • MDL number MFCD30489750
  • Cas Number : 2029049-79-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.ambeed.com/products/2029049-79-2.html

Safety Information

MSDS https://www.ambeed.com/products/2029049-79-2.html
Availability details for
Quantity
  • Availability
  • Add to Cart