AMBH5806470B Ambeed, Inc.

2-(4-Methoxybenzylidene)imino-2-deoxy-1,3,4,6-Tetra-O-acetyl-�D-glucopyranose

  • MDL number MFCD10487691
  • Cas Number : 7597-81-1
  • Molecular Weight : 465.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.ambeed.com/products/7597-81-1.html

Safety Information

MSDS https://www.ambeed.com/products/7597-81-1.html
Availability details for
Quantity
  • Availability
  • Add to Cart