AMBH5806470C Ambeed, Inc.

Methyl benzofuran-2-carboxylate

  • MDL number MFCD01159618
  • Cas Number : 1646-27-1
  • Molecular Weight : 176.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 257.7�t 760 mmHg
Iupac Name methyl 1-benzofuran-2-carboxylate
Inchi Code 1S/C10H8O3/c1-12-10(11)9-6-7-4-2-3-5-8(7)13-9/h2-6H,1H3
Inchi Key GIJIKNIHYNIDSJ-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 257.7�t 760 mmHg
Product Link https://www.ambeed.com/products/1646-27-1.html

Safety Information

signalword Warning
pcodes P261-P264-P270-P271-P280-P301+P312+P330-P302+P352+P312-P304+P340+P312-P363-P501
MSDS https://www.ambeed.com/products/1646-27-1.html
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart