AMBH58064713 Ambeed, Inc.

6-Nitrophthalazine

  • MDL number MFCD00475919
  • Cas Number : 120493-12-1
  • Molecular Weight : 175.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/120493-12-1.html

Safety Information

MSDS https://www.ambeed.com/products/120493-12-1.html
Availability details for
Quantity
  • Availability
  • Add to Cart