AMBH58064714 Ambeed, Inc.

(S)-2-(2-(Diphenylphosphino)phenyl)-4-isopropyl-4,5-dihydrooxazole

  • MDL number MFCD03427599
  • Cas Number : 148461-14-7
  • Molecular Weight : 373.4272

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,Store in freezer, under -20
Physical Form solid
Shipping Temperature Inert atmosphere,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/148461-14-7.html

Safety Information

signalword Warning
pcodes P261-P280-P301+P312-P302+P352-P305+P351+P338
MSDS https://www.ambeed.com/products/148461-14-7.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart