AMBH5806471D Ambeed, Inc.

1-(2-Bromopyridin-4-yl)cyclopropanecarbonitrile

  • MDL number MFCD20278074
  • Cas Number : 1279815-46-1
  • Molecular Weight : 223.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(2-bromo-4-pyridinyl)cyclopropanecarbonitrile
Inchi Code 1S/C9H7BrN2/c10-8-5-7(1-4-12-8)9(6-11)2-3-9/h1,4-5H,2-3H2
Inchi Key HKLRZPZCPJJBTP-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/1279815-46-1.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/1279815-46-1.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart