AMBH58064722 Ambeed, Inc.

Ethyl 2-(1H-benzo[d]imidazol-2-yl)acetate

  • MDL number MFCD00624872
  • Cas Number : 14741-71-0
  • Molecular Weight : 204.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 408.3�t 760 mmHg
Iupac Name ethyl 1H-benzimidazol-2-ylacetate
Inchi Code 1S/C11H12N2O2/c1-2-15-11(14)7-10-12-8-5-3-4-6-9(8)13-10/h3-6H,2,7H2,1H3,(H,12,13)
Inchi Key JTVPWPDLKUQDAU-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 408.3�t 760 mmHg
Product Link https://www.ambeed.com/products/14741-71-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/14741-71-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart