AMBH58064725 Ambeed, Inc.

4-Isopropyl-2,2-dimethylbenzo[d][1,3]dioxole

  • MDL number MFCD12024638
  • Cas Number : 201166-22-5
  • Molecular Weight : 192.2542

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,Store in freezer, under -20
Physical Form liquid
Shipping Temperature Inert atmosphere,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/201166-22-5.html

Safety Information

signalword Danger
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/201166-22-5.html
pictograms GHS02,GHS07
hstatements H226-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart