AMBH58064738 Ambeed, Inc.

1-(4-Aminopiperidin-1-yl)-2,2,2-trifluoroethanone

  • MDL number MFCD11849675
  • Cas Number : 497177-66-9
  • Molecular Weight : 196.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/497177-66-9.html

Safety Information

signalword Warning
pcodes P305+P351+P338
MSDS https://www.ambeed.com/products/497177-66-9.html
pictograms GHS07
hstatements H302-H319
Availability details for
Quantity
  • Availability
  • Add to Cart