AMBH5806473B Ambeed, Inc.

Imeglimin hydrochloride

  • MDL number MFCD28167741
  • Cas Number : 775351-61-6
  • Molecular Weight : 191.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.ambeed.com/products/775351-61-6.html

Safety Information

MSDS https://www.ambeed.com/products/775351-61-6.html
Availability details for
Quantity
  • Availability
  • Add to Cart