AMBH5806473D Ambeed, Inc.

(S)-Methyl 2,3-diaminopropanoate dihydrochloride

  • MDL number MFCD09998031
  • Cas Number : 147857-43-0
  • Molecular Weight : 191.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.ambeed.com/products/147857-43-0.html

Safety Information

MSDS https://www.ambeed.com/products/147857-43-0.html
Availability details for
Quantity
  • Availability
  • Add to Cart