AMBH58064742 Ambeed, Inc.

1-Benzyl-1,4-dihydronicotinamide

  • MDL number MFCD00059769
  • Cas Number : 952-92-1
  • Molecular Weight : 214.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 433.4�t 760 mmHg
Iupac Name 1-benzyl-1,4-dihydro-3-pyridinecarboxamide
Inchi Code 1S/C13H14N2O/c14-13(16)12-7-4-8-15(10-12)9-11-5-2-1-3-6-11/h1-6,8,10H,7,9H2,(H2,14,16)
Inchi Key CMNUYDSETOTBDE-UHFFFAOYSA-N

Description

Purity 95%
Flash Point 215.9
Storage temp. Sealed in dry,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 433.4�t 760 mmHg
Product Link https://www.ambeed.com/products/952-92-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/952-92-1.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart