AMBH58064746 Ambeed, Inc.

2-Ethyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

  • MDL number MFCD19441153
  • Cas Number : 82954-89-0
  • Molecular Weight : 156.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/82954-89-0.html

Safety Information

signalword Danger
pcodes P210-P233-P240-P241+P242+P243-P264-P280-P302+P352+P332+P313+P362+P364-P305+P351+P338+P337+P313-P370+P378-P403+P235-P501
MSDS https://www.ambeed.com/products/82954-89-0.html
pictograms GHS02,GHS07
hstatements H226-H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart