AMBH58064751 Ambeed, Inc.

2-((2-Acetamido-6-oxo-1H-purin-9(6H)-yl)methoxy)ethyl acetate

  • MDL number MFCD00267706
  • Cas Number : 75128-73-3
  • Molecular Weight : 309.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 204°C at N/A mmHg
Purity 97
Storage temp. Sealed in dry,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/75128-73-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/75128-73-3.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart