AMBH58064755 Ambeed, Inc.

[6-Methoxy-5-(trifluoromethyl)pyridin-3-yl]boronic acid

  • MDL number MFCD18254530
  • Cas Number : 1420830-61-0
  • Molecular Weight : 220.9416

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1420830-61-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1420830-61-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart