AMBH58064758 Ambeed, Inc.

3-Amino-2-fluorobenzonitrile

  • MDL number MFCD08062434
  • Cas Number : 873697-68-8
  • Molecular Weight : 136.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Keep in dark place,Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/873697-68-8.html

Safety Information

signalword Danger
pcodes P261-P280-P301+P310-P311
MSDS https://www.ambeed.com/products/873697-68-8.html
pictograms GHS06
hstatements H301+H311+H331
Availability details for
Quantity
  • Availability
  • Add to Cart