AMBH5806475A Ambeed, Inc.

ILK-IN-2

  • MDL number MFCD29924730
  • Cas Number : 2070015-22-2
  • Molecular Weight : 533.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N-Methyl-3-(1-(4-(piperazin-1-yl)phenyl)-3-(4'-(trifluoromethyl)-[1,1'-biphenyl]-4-yl)-1H-pyrazol-5-yl)propanamide
Purity 98
Storage temp. Keep in dark place,Sealed in dry,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/2070015-22-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/2070015-22-2.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart