AMBH58064762 Ambeed, Inc.

(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(1-((benzyloxy)methyl)-1H-imidazol-5-yl)propanoic acid

  • MDL number MFCD00077059
  • Cas Number : 84891-19-0
  • Molecular Weight : 497.5418

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Store in freezer, under -20
Physical Form solid
Shipping Temperature Sealed in dry,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/84891-19-0.html

Safety Information

signalword Warning
pcodes P261-P280-P301+P312-P302+P352-P305+P351+P338
MSDS https://www.ambeed.com/products/84891-19-0.html
pictograms GHS07
hstatements H302-H315-H320-H335
Availability details for
Quantity
  • Availability
  • Add to Cart