AMBH58064765 Ambeed, Inc.

4-(4-Nitrophenyl)butan-2-one

  • MDL number MFCD11655141
  • Cas Number : 30780-19-9
  • Molecular Weight : 193.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(4-nitrophenyl)-2-butanone
Inchi Code 1S/C10H11NO3/c1-8(12)2-3-9-4-6-10(7-5-9)11(13)14/h4-7H,2-3H2,1H3
Inchi Key BQCIKEHCNRRAKU-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/30780-19-9.html

Safety Information

signalword Warning
pcodes P261-P280-P301+P312-P302+P352-P305+P351+P338
MSDS https://www.ambeed.com/products/30780-19-9.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart