AMBH58064769 Ambeed, Inc.

3-((1,3-Dioxoisoindolin-2-yl)methyl)benzoic acid

  • MDL number MFCD00418210
  • Cas Number : 106352-01-6
  • Molecular Weight : 281.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 511.4�t 760 mmHg
Iupac Name 3-[(1,3-dioxo-1,3-dihydro-2H-isoindol-2-yl)methyl]benzoic acid
Inchi Code 1S/C16H11NO4/c18-14-12-6-1-2-7-13(12)15(19)17(14)9-10-4-3-5-11(8-10)16(20)21/h1-8H,9H2,(H,20,21)
Inchi Key ZUWJKVVWOLYFRK-UHFFFAOYSA-N

Description

Purity 95%
Flash Point 263.1
Storage temp. Inert atmosphere,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 511.4�t 760 mmHg
Product Link https://www.ambeed.com/products/106352-01-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/106352-01-6.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart