AMBH5806476B Ambeed, Inc.

4-Methyl-1-piperazinecarbonyl Chloride Hydrochloride

  • MDL number MFCD01074875
  • Cas Number : 55112-42-0
  • Molecular Weight : 199.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/55112-42-0.html

Safety Information

signalword Danger
pcodes P280-P305+P351+P338-P310
MSDS https://www.ambeed.com/products/55112-42-0.html
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart