AMBH58064770 Ambeed, Inc.

Silver(I) chloride

  • MDL number MFCD00003399
  • Cas Number : 7783-90-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Room Temp
Physical Form solid
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.ambeed.com/products/7783-90-6.html

Safety Information

MSDS https://www.ambeed.com/products/7783-90-6.html
Availability details for
Quantity
  • Availability
  • Add to Cart