AMBH58064778 Ambeed, Inc.

Ethyl (2-chloroethyl)carbamate

  • MDL number MFCD04038117
  • Cas Number : 6329-26-6
  • Molecular Weight : 151.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 225.9�t 760 mmHg
Iupac Name ethyl 2-chloroethylcarbamate
Inchi Code 1S/C5H10ClNO2/c1-2-9-5(8)7-4-3-6/h2-4H2,1H3,(H,7,8)
Inchi Key QHSHDVYEJKLXLB-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Sealed in dry,Store in freezer, under -20
Physical Form Colorless to Pale-yellow to Yellow-brown Sticky Oil to Semi-Solid or Liquid
Shipping Temperature Shipped in cold pack
Country of Origin CN
Boiling point 225.9�t 760 mmHg
Product Link https://www.ambeed.com/products/6329-26-6.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/6329-26-6.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart