AMBH5806477D Ambeed, Inc.

2-Amino-6-iodobenzaldehyde

  • MDL number MFCD17214948
  • Cas Number : 1261872-97-2
  • Molecular Weight : 247.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.ambeed.com/products/1261872-97-2.html

Safety Information

MSDS https://www.ambeed.com/products/1261872-97-2.html
Availability details for
Quantity
  • Availability
  • Add to Cart