AMBH5806478B Ambeed, Inc.

3-Bromo-6-oxo-1,6-dihydropyridine-2-carbonitrile

  • MDL number MFCD20482850
  • Cas Number : 1186637-40-0
  • Molecular Weight : 199.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Bromo-6-hydroxypicolinonitrile
Purity 95
Storage temp. Inert atmosphere,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/1186637-40-0.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/1186637-40-0.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart