AMBH58064794 Ambeed, Inc.

2-(1H-Pyrrolo[2,3-b]pyridin-3-yl)ethanamine

  • MDL number MFCD10697517
  • Cas Number : 4649-12-1
  • Molecular Weight : 161.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Flash Point 197.9
Storage temp. Keep in dark place,Inert atmosphere,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 358.4°C at 760 mmHg
Product Link https://www.ambeed.com/products/4649-12-1.html

Safety Information

signalword Danger
pcodes P280-P305+P351+P338-P310
hcodes 8
MSDS https://www.ambeed.com/products/4649-12-1.html
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart