AMBH58064796 Ambeed, Inc.

Lactobionic acid

  • MDL number MFCD00078147
  • Cas Number : 96-82-2
  • Molecular Weight : 358.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.ambeed.com/products/96-82-2.html

Safety Information

MSDS https://www.ambeed.com/products/96-82-2.html
Availability details for
Quantity
  • Availability
  • Add to Cart