AMBH5806479A Ambeed, Inc.

(4-Chlorophenyl)trimethoxysilane

  • MDL number MFCD29037385
  • Cas Number : 35692-30-9
  • Molecular Weight : 232.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-chlorophenyl)trimethoxysilane
Inchi Code 1S/C9H13ClO3Si/c1-11-14(12-2,13-3)9-6-4-8(10)5-7-9/h4-7H,1-3H3
Inchi Key SWYZNMNIUXTWTF-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Keep in dark place,Inert atmosphere,2-8
Physical Form liquid
Shipping Temperature Cold-chain transportation
Country of Origin CN
Product Link https://www.ambeed.com/products/35692-30-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/35692-30-9.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart