AMBH5806479C Ambeed, Inc.

(+)-Pulegone

  • MDL number MFCD00063000
  • Cas Number : 89-82-7
  • Molecular Weight : 152.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 92
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/89-82-7.html

Safety Information

signalword Danger
pcodes P273-P301+P312+P330
MSDS https://www.ambeed.com/products/89-82-7.html
pictograms GHS07,GHS08
hstatements H302-H351-H412
Availability details for
Quantity
  • Availability
  • Add to Cart