AMBH5806479D Ambeed, Inc.

tert-Butyl 2-oxomorpholine-4-carboxylate

  • MDL number MFCD28098563
  • Cas Number : 1140502-97-1
  • Molecular Weight : 201.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 2-oxomorpholine-4-carboxylate
Inchi Code 1S/C9H15NO4/c1-9(2,3)14-8(12)10-4-5-13-7(11)6-10/h4-6H2,1-3H3
Inchi Key PDZLTNQMFWCEJQ-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. Sealed in dry,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/1140502-97-1.html

Safety Information

signalword Warning
pcodes P264-P270-P301+P312-P330
MSDS https://www.ambeed.com/products/1140502-97-1.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart