AMBH5806479E Ambeed, Inc.

5,6-Difluoropicolinic acid

  • MDL number MFCD09031175
  • Cas Number : 851386-38-4
  • Molecular Weight : 159.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 851386-38-4
bp 291.323�t 760 mmHg
Iupac Name 5,6-difluoro-2-pyridinecarboxylic acid
Inchi Code 1S/C6H3F2NO2/c7-3-1-2-4(6(10)11)9-5(3)8/h1-2H,(H,10,11)
Inchi Key NWMYBARTEFULTJ-UHFFFAOYSA-N

Description

Melting Point 851386-38-4
Purity 95%
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 291.323�t 760 mmHg
Product Link https://www.ambeed.com/products/851386-38-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/851386-38-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart