AMBH580647AA Ambeed, Inc.

Benzo[c][1,2,5]thiadiazol-5-ol

  • MDL number MFCD00457955
  • Cas Number : 768-10-5
  • Molecular Weight : 152.1738

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Physical Form solid
Shipping Temperature Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/768-10-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/768-10-5.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart