AMBH580647AB Ambeed, Inc.

1-(4-Fluorobenzyl)-6-oxo-1,6-dihydropyridazine-3-carboxylic acid

  • MDL number MFCD16652636
  • Cas Number : 1105192-19-5
  • Molecular Weight : 248.2099

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1105192-19-5.html

Safety Information

signalword Warning
pcodes P233-P260-P261-P264-P271-P280-P302+P352-P304-P304+P340-P305+P351+P338-P312-P321-P332+P313-P337+P313-P340-P362-P403-P403+P233-P405-P501
MSDS https://www.ambeed.com/products/1105192-19-5.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart