AMBH580647B0 Ambeed, Inc.

Belotecan HCl

  • MDL number MFCD07772313
  • Cas Number : 213819-48-8
  • Molecular Weight : 469.9605

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/213819-48-8.html

Safety Information

signalword Warning
pcodes P305+P351+P338-P280
MSDS https://www.ambeed.com/products/213819-48-8.html
pictograms GHS07
hstatements H317-H319
Availability details for
Quantity
  • Availability
  • Add to Cart