AMBH580647B6 Ambeed, Inc.

3-Butyl-1-methyl-1H-imidazol-3-ium hydrogensulfate

  • MDL number MFCD06798197
  • Cas Number : 262297-13-2
  • Molecular Weight : 236.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/262297-13-2.html

Safety Information

signalword Danger
pcodes P280-P305+P351+P338-P310
MSDS https://www.ambeed.com/products/262297-13-2.html
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart