AMBH580647B9 Ambeed, Inc.

Methyl 2,3,4-Tri-O-acetyl-b-D-glucuronic Acid Methyl Ester

  • MDL number N/A
  • Cas Number : 34213-34-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.ambeed.com/products/34213-34-8.html

Safety Information

MSDS https://www.ambeed.com/products/34213-34-8.html
Availability details for
Quantity
  • Availability
  • Add to Cart