AMBH580647C4 Ambeed, Inc.

Phosphocholine Chloride Calcium Salt Tetrahydrate

  • MDL number MFCD00011755
  • Cas Number : 4826-71-5
  • Molecular Weight : 257.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/4826-71-5.html

Safety Information

signalword Danger
pcodes P201-P261-P280-P305+P351+P338-P308+P313
MSDS https://www.ambeed.com/products/4826-71-5.html
pictograms GHS08
hstatements H315-H317-H319-H335-H360
Availability details for
Quantity
  • Availability
  • Add to Cart