AMBH580647C8 Ambeed, Inc.

6-Chloro-2-(trifluoromethyl)-3H-imidazo[4,5-b]pyridine

  • MDL number MFCD00223328
  • Cas Number : 13577-71-4
  • Molecular Weight : 221.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.ambeed.com/products/13577-71-4.html

Safety Information

MSDS https://www.ambeed.com/products/13577-71-4.html
Availability details for
Quantity
  • Availability
  • Add to Cart