AMBH580647C9 Ambeed, Inc.

Benzyl 2-(hydroxymethyl)piperazine-1-carboxylate

  • MDL number MFCD09954310
  • Cas Number : 1131595-00-0
  • Molecular Weight : 250.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name benzyl 2-(hydroxymethyl)-1-piperazinecarboxylate
Inchi Code 1S/C13H18N2O3/c16-9-12-8-14-6-7-15(12)13(17)18-10-11-4-2-1-3-5-11/h1-5,12,14,16H,6-10H2
Inchi Key DUSWOUMCIHZHBH-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Keep in dark place,Sealed in dry,2-8
Physical Form Liquid or solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/1131595-00-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1131595-00-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart