AMBH580647CF Ambeed, Inc.

2-Bromo-5-nitro-1,3,4-thiadiazole

  • MDL number MFCD12923090
  • Cas Number : 22758-10-7
  • Molecular Weight : 210.0093

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/22758-10-7.html

Safety Information

signalword Warning
pcodes P261-P280-P301+P312-P302+P352-P305+P351+P338
MSDS https://www.ambeed.com/products/22758-10-7.html
pictograms GHS07
hstatements H302-H315-H320-H335
Availability details for
Quantity
  • Availability
  • Add to Cart