AMBH580647D0 Ambeed, Inc.

Vinorelbine Ditartrate

  • MDL number MFCD03613607
  • Cas Number : 125317-39-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Physical Form solid
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.ambeed.com/products/125317-39-7.html

Safety Information

MSDS https://www.ambeed.com/products/125317-39-7.html
Availability details for
Quantity
  • Availability
  • Add to Cart