AMBH580647D7 Ambeed, Inc.

tert-Butyl (4-iodobutyl)carbamate

  • MDL number MFCD09951820
  • Cas Number : 262278-40-0
  • Molecular Weight : 299.1492

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Inert atmosphere,Store in freezer, under -20
Physical Form liquid
Shipping Temperature Keep in dark place,Inert atmosphere,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/262278-40-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/262278-40-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart