AMBH580647E1 Ambeed, Inc.

3-Oxo-3-(piperidin-1-yl)propanenitrile

  • MDL number MFCD00006487
  • Cas Number : 15029-30-8
  • Molecular Weight : 152.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/15029-30-8.html

Safety Information

MSDS https://www.ambeed.com/products/15029-30-8.html
Availability details for
Quantity
  • Availability
  • Add to Cart