AMBH580647E3 Ambeed, Inc.

6-Chloro-2,4-dimethylpyridazin-3(2H)-one

  • MDL number MFCD27923629
  • Cas Number : 1114563-58-4
  • Molecular Weight : 158.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-chloro-2,4-dimethylpyridazin-3(2H)-one
Inchi Code 1S/C6H7ClN2O/c1-4-3-5(7)8-9(2)6(4)10/h3H,1-2H3
Inchi Key BOVMTTYMJWYEIV-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/1114563-58-4.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/1114563-58-4.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart